R&D Systems
Manufacturer Code:AF3904SP
Catalog # AF3904SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MuSK Glu22-Ser503 Accession # O15146 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 MGC126323 MGC126324 muscle skeletal receptor tyrosine kinase MuSK skeletal receptor tyrosine-protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK
Gene Aliases: CMS9; FADS; MUSK
UniProt ID: (Human) O15146
Entrez Gene ID: (Human) 4593
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.