R&D Systems
Manufacturer Code:AF2284SP
Catalog # AF2284SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Mre11 Glu423-Arg708 Accession # P49959 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: AT-like disease; ATLDmeiotic recombination (S. cerevisiae) 11 homolog A AT-like disease DNA recombination and repair protein double-strand break repair protein MRE11A endo/exonuclease Mre11 HNGS1 Meiotic recombination 11 homolog 1 Meiotic recombination 11 homolog A MRE11 homolog 1 MRE11 homolog A MRE11 meiotic recombination 11 homolog A (S. cerevisiae) MRE11A MRE11B; DNA recombination and repair protein; Double-strand break repair protein MRE11; Double-strand break repair protein MRE11A; endo/exonuclease Mre11; Meiotic recombination 11 homolog 1; Meiotic recombination 11 homolog A; MRE11 double strand break repair nuclease A; MRE11 homolog 1; MRE11 homolog A; MRE11 homolog, double strand break repair nuclease A; MRE11 meiotic recombination 11 homolog A; MRE11 meiotic recombination 11-like protein A
Gene Aliases: ATLD; HNGS1; MRE11; MRE11A; MRE11B
UniProt ID: (Human) P49959
Entrez Gene ID: (Human) 4361
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.