Southern Biotechnology
Manufacturer Code:615832
Catalog # OB615832
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Goat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Secondary Antibody |
Clone | SB115d |
Immunogen | Pooled goat IgG |
Conjugate | Alexa Fluor 555 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C Protect from light |
Target | IgG2a |
Mouse Monoclonal Secondary Antibody
For Research Use Only
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.