Invitrogen
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | 5 µL/1x10^6 cells |
Host / Isotype | Mouse / IgG1 |
Class | Monoclonal |
Type | Secondary Antibody |
Clone | CH2 |
Immunogen | Purified human IgM |
Conjugate | Alexa Fluor™ 647 |
Form | Liquid |
Concentration | 0.08 mg/mL |
Storage conditions | 4° C, store in dark |
RRID | AB_2633296 |
Target | IgM |
Antibody Form | Whole Antibody |
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.