Novus Biologicals
Manufacturer Code:NBP128241AF647
Catalog # NBP128241Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 74-22-15 |
Immunogen | Fresh dd miniature swine thymocytes |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (Ala-IL-8)77; (Ser-IL-8)72; 3-10C AMCF-I C-X-C motif chemokine 8 CXCL8SCYB8 Emoctakin GCP-1TSG-1 interleukin 8 K60 LECT MDNCFb-ENAP member 8 MONAPGCP1 NAP-1NAP1 Neutrophil-activating protein 1 Protein 3-10C T cell chemotactic factor T-cell chemotactic factor; alveolar macrophage chemotactic factor I; beta endothelial cell-derived neutrophil activating peptide; beta-thromboglobulin-like protein; C-X-C motif chemokine 8; Chemokine (C-X-C motif) ligand 8; Emoctakin; GCP-1; GCP/IL-8 protein I; GCP/IL-8 protein II; GCP/IL-8 protein III; GCP/IL-8 protein IV; GCP/IL-8 protein V; GCP/IL-8 protein VI; Granulocyte chemotactic protein 1; IL-8; IL-8(1-77); IL-8(5-77); IL-8(6-77); IL-8(7-77); IL-8(8-77); IL-8(9-77); IL8/NAP1 form I; IL8/NAP1 form II; IL8/NAP1 form III; IL8/NAP1 form IV; IL8/NAP1 form V; IL8/NAP1 form VI; interleukin 8; Interleukin-8; lung giant cell carcinoma-derived chemotactic protein; lymphocyte derived neutrophil activating peptide; Lymphocyte-derived neutrophil-activating factor; LYNAP; MDNCF; MDNCF-a; MDNCF-b; MDNCF-c; MONAP; Monocyte-derived neutrophil chemotactic factor; Monocyte-derived neutrophil-activating peptide; NAF; NAP-1; Neutrophil-activating factor; neutrophil-activating peptide 1; Neutrophil-activating protein 1; Protein 3-10C; small inducible cytokine subfamily B, member 8; T-cell chemotactic factor; tumor necrosis factor-induced gene 1
Gene Aliases: CXCL8; GCP-1; GCP1; IL8; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP-1; NAP1
UniProt ID: (Human) P10145
Entrez Gene ID: (Human) 3576
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.