Invitrogen
Manufacturer Code:MA535138
Catalog # PIMA535138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0641 |
Immunogen | A synthesized peptide derived from human Monoamine Oxidase B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 6330414K01Rik Amine oxidase [flavin-containing] B BOS_26102 HGNC:6834 MAO b Maob MAO-B MGC26382 monoamine oxidase B Monoamine oxidase type B RP1-201D17__B.1 tyramine oxidase; adrenalin oxidase; Amine oxidase [flavin-containing] B; MAO, brain; MAO, platelet; MAO-B; Monoamine oxidase type B; tyramine oxidase
Gene Aliases: MAOB
UniProt ID: (Human) P27338
Entrez Gene ID: (Human) 4129
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.