Invitrogen
Manufacturer Code:PA582012
Catalog # PIPA582012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human MAOB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenalin oxidase; Amine oxidase [flavin-containing] B; MAO, brain; MAO, platelet; MAO-B; Monoamine oxidase type B; monoamine oxidase-B; tyramine oxidase
Gene Aliases: 6330414K01Rik; MAO-B; MAOB
UniProt ID: (Human) P27338, (Mouse) Q8BW75, (Rat) P19643
Entrez Gene ID: (Human) 4129, (Mouse) 109731, (Rat) 25750
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.