Invitrogen
Manufacturer Code:PA542382
Catalog # PIPA542382
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human MAOB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenalin oxidase; Amine oxidase [flavin-containing] B; MAO, brain; MAO, platelet; MAO-B; Monoamine oxidase B MGC26382; Monoamine oxidase type B; monoamine oxidase-B; tyramine oxidase
Gene Aliases: 6330414K01Rik; MAO-B; MAOB
UniProt ID: (Bovine) P56560, (Human) P27338, (Pig) Q6PLK3, (Dog) Q7YRB7, (Mouse) Q8BW75, (Guinea pig) P58028, (Rat) P19643
Entrez Gene ID: (Rabbit) 100357007, (Bovine) 338445, (Horse) 100033868, (Human) 4129, (Pig) 414909, (Dog) 403451, (Mouse) 109731, (Guinea pig) 100135573, (Rat) 25750
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.