Invitrogen
Manufacturer Code:PA528338
Catalog # PIPA528338
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 163 and 501 of Human Monoamine Oxidase B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenalin oxidase; Amine oxidase [flavin-containing] B; MAO brain MAO platelet MAO-B adrenalin oxidase amine oxidase &lsqbflavin-containing] B monoamine oxidase type B tyramine oxidase; MAO, brain; MAO, platelet; MAO-B; Monoamine oxidase type B; tyramine oxidase
Gene Aliases: 6330414K01Rik; MAO-B; MAOB
UniProt ID: (Human) P27338, (Mouse) Q8BW75
Entrez Gene ID: (Human) 4129, (Mouse) 109731
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.