Novus Biologicals
Manufacturer Code:NBP187493
Catalog # NBP187493
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:WRTMDDMGREIPSDAPWKAPLAEEWDNMTMKELLDKLCWTESAKQLATLFVNLCVTAETHEVSALWFLWYVKQCGGTTRIISTTNGGQERKFVGGSGQVSERIMDLLGDRVKLERPVIYIDQTRENVLVETLNHEMYEAKYVISAIPPTLGAPHR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adrenalin oxidase; adrenalin oxidase amine oxidase [flavin-containing] B EC 1.4.3 EC 1.4.3.4 MAO brain MAO platelet MAO-B MGC26382 monoamine oxidase B Monoamine oxidase type B tyramine oxidase; Amine oxidase [flavin-containing] B; MAO, brain; MAO, platelet; MAO-B; Monoamine oxidase type B; tyramine oxidase
Gene Aliases: MAOB
UniProt ID: (Human) P27338
Entrez Gene ID: (Human) 4129
Molecular Function:
DNA binding protein
DNA methyltransferase
methyltransferase
nucleic acid binding
oxidase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.