Invitrogen
Manufacturer Code:PA570605
Catalog # PIPA570605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human Moesin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 70; Membrane-organizing extension spike protein; Moesin
Gene Aliases: C78546; HEL70; MSN
UniProt ID: (Bovine) Q2HJ49, (Human) P26038, (Rat) O35763, (Mouse) P26041
Entrez Gene ID: (Bovine) 540426, (Horse) 100065348, (Dog) 491924, (Rabbit) 100338273, (Human) 4478, (Rat) 81521, (Guinea pig) 100735797, (Mouse) 17698
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.