Invitrogen
Manufacturer Code:MA514928
Catalog # PIMA514928
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | E.611.4 |
Immunogen | Synthetic peptide corresponding to the sequence of human Mnk1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: MAP kinase interacting kinase 1 MAP kinase signal-integrating kinase 1 MAP kinase-interacting serine/threonine kinase 1 MAP kinase-interacting serine/threonine-protein kinase 1 OTTHUMP00000009675 OTTHUMP00000009676; MAP kinase signal-integrating kinase 1; MAP kinase-interacting serine/threonine-protein kinase 1; MAPK signal-integrating kinase 1
Gene Aliases: 2410048M24Rik; MKNK1; MNK1
UniProt ID: (Human) Q9BUB5, (Mouse) O08605
Entrez Gene ID: (Human) 8569, (Mouse) 17346
Molecular Function:
cytoskeletal protein
kinase
microtubule family cytoskeletal protein
non-motor microtubule binding protein
non-receptor serine/threonine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.