Abnova
Manufacturer Code:MAB18209
Catalog # 89-338-134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S51-82 |
Immunogen | Recombinant protein corresponding to full length rat Mmp9. |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: -; 92 kDa gelatinase; 92 kDa type IV collagenase; 92-kDa type IV collagenase; Gelatinase B; GELB; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); Matrix metalloproteinase-9; MMP-9
Gene Aliases: Mmp9
UniProt ID: (Rat) P50282
Entrez Gene ID: (Rat) 81687
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.