Abnova
Manufacturer Code:PAB8850
Catalog # 89-133-976
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to rat Mmp9. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against a recombinant Mmp9.
For Research Use Only
Protein Aliases: 92 kDa gelatinase; 92 kDa type IV collagenase; 92-kDa type IV collagenase; Gelatinase B; GELB; matrix metalloproteinase 9 (gelatinase B 92-kDa type IV collagenase); matrix metalloproteinase 9 (gelatinase B, 92-kDa type IV collagenase); Matrix metalloproteinase-9; MMP-9
Gene Aliases: Mmp9
UniProt ID: (Rat) P50282
Entrez Gene ID: (Rat) 81687
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.