Proteintech
Manufacturer Code:264691AP150UL
Catalog # 50-173-5350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mitoferrin 1 Fusion Protein Ag24876 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MFRN Mfrn1 Mitochondrial iron transporter 1 Mitoferrin 1 SLC25A37; Mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitoferrin; mitoferrin 1; Mitoferrin-1; predicted protein of HQ2217; solute carrier family 25 (mitochondrial iron transporter), member 37; Solute carrier family 25 member 37; solute carrier family 25, member 37
Gene Aliases: 1700020E22Rik; 4930513O14Rik; 4930526G11Rik; AI848481; C330015G08Rik; frascati; HT015; MFRN; MFRN1; mitoferrin; MSC; MSCP; PRO1278; PRO1584; PRO2217; SLC25A37
UniProt ID: (Human) Q9NYZ2, (Mouse) Q920G8
Entrez Gene ID: (Human) 51312, (Mouse) 67712
Molecular Function:
RNA binding protein
amino acid transporter
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
mitochondrial carrier protein
nucleic acid binding
ribosomal protein
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.