Invitrogen
Manufacturer Code:MA516214
Catalog # PIMA516214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 21D1418 |
Immunogen | Synthetic peptide corresponding to amino acids 301-312 (PSTGLCSPDLVN) of MITF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHe32; black eyed white; Class E basic helix-loop-helix protein 32; MI WS2 WS2A bHLHe32 class E basic helix-loop-helix protein 32; microphtalmia-associated transcription factor; Microphthalmia-associated transcription factor
Gene Aliases: BCC2; BHLHE32; Bw; CMM8; Gsfbcc2; MI; MITF; Vit; Vitiligo; Wh; WS2; WS2A
UniProt ID: (Human) O75030, (Rat) O88368, (Mouse) Q08874
Entrez Gene ID: (Human) 4286, (Rat) 25094, (Mouse) 17342
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.