Novus Biologicals
Manufacturer Code:NBP152031
Catalog # NBP152031
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-RKNQYVSNGLRDFA corresponding to internal region according to NP_036577.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Differentially expressed in cancerous and non-cancerous lung cells 1; Differentially expressed in cancerous and non-cancerous lung cells 1 DIL-1 DIL1FLJ34460 DKFZp686G21139 FLJ16313 FLJ22401 MINDIN M-SPONDIN spondin 2 extracellular matrix protein spondin-2; DIL-1; Mindin; spondin 2, extracellular matrix protein; Spondin-2
Gene Aliases: DIL-1; DIL1; M-SPONDIN; MINDIN; SPON2; UNQ435/PRO866
UniProt ID: (Human) Q9BUD6
Entrez Gene ID: (Human) 10417
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.