R&D Systems
Manufacturer Code:AF2660
Catalog # AF2660
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Mimecan Ala20-Phe298 Accession # P20774 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: corneal keratan sulfate proteoglycan; DKFZp586P2421 KSPG25 mimecan mimecan proteoglycan OGN OIF OIFOG osteoglycin osteoglycin (osteoinductive factor) Osteoinductive factor SLRR3A SLRR3Acorneal keratan sulfate proteoglycan; Mimecan; mimecan proteoglycan; OIF; Osteoglycin; Osteoinductive factor
Gene Aliases: OG; OGN; OIF; SLRR3A
UniProt ID: (Human) P20774
Entrez Gene ID: (Human) 4969
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.