Invitrogen
Manufacturer Code:PA588982
Catalog # PIPA588982
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 223-462 of human ERRFI1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ERBB receptor feedback inhibitor 1; Gene 33 polypeptide; MIG-6; Mitogen-inducible gene 6 protein homolog; RALT; Receptor-associated late transducer
Gene Aliases: 33; Errfi1; gene 33; Mig6; Ralt; RGD1307599; Xxx
UniProt ID: (Rat) P05432
Entrez Gene ID: (Rat) 313729
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.