Invitrogen
Manufacturer Code:PA5110930
Catalog # PIPA5110930
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human Mig-6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300002F13Rik 33 AI788755 ERBB receptor feedback inhibitor 1 ERRFI1 gene 33 Gene 33 polypeptide GENE-33 Mig6 MIG-6 Mitogen-inducible gene 6 protein Mitogen-inducible gene 6 protein homolog RALT Receptor-associated late transducer RGD1307599 Xxx; ERBB receptor feedback inhibitor 1; MIG-6; Mitogen-inducible gene 6 protein; receptor-associated late transducer
Gene Aliases: ERRFI1; GENE-33; MIG-6; MIG6; RALT
UniProt ID: (Human) Q9UJM3
Entrez Gene ID: (Human) 54206
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.