R&D Systems
Manufacturer Code:AF258PB
Catalog # AF258PB
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Midkine Lys23-Asp143 Accession # P21741 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Amphiregulin-associated protein; Amphiregulin-associated protein ARAP MDK MEK Midgestation and kidney protein midkine midkine (neurite growth-promoting factor 2) MK1 MKARAP NEGF2 NEGF2FLJ27379 Neurite outgrowth-promoting factor 2 Neurite outgrowth-promoting protein; ARAP; Midgestation and kidney protein; Midkine; MK; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; retinoic acid inducible factor
Gene Aliases: ARAP; MDK; MK; MK1; NEGF2
UniProt ID: (Human) P21741
Entrez Gene ID: (Human) 4192
Molecular Function: cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.