Invitrogen
Manufacturer Code:MA514154
Catalog # PIMA514154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Gel Shift (GS) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | D5 |
Immunogen | N-terminal fragment of human Mi protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHe32; Class E basic helix-loop-helix protein 32; melanogenesis associated transcription factor; microphtalmia-associated transcription factor; Microphthalmia-associated transcription factor; MITF-H
Gene Aliases: BHLHE32; CMM8; MI; MITF; MITF-H; MITF-M; WS2; WS2A
UniProt ID: (Human) O75030
Entrez Gene ID: (Dog) 415126, (Human) 4286
Molecular Function:
basic helix-loop-helix transcription factor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.