Proteintech
Manufacturer Code:107991AP150UL
Catalog # 50-172-6608
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mhc Fusion Protein Ag1254 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta-myosin heavy chain; Bsh CG17927 Dm II DmelCG17927 DmMHC DROMHC DroMII FBgn0002741 Ifm(2)2 l(2)36Ae l(2)k10423 l(2)M66 Mhc Mhc36B mMHC MRP Myo Myosin heavy chain Nup Sht sMHC Stp; CG17927-PA; CG17927-PB; CG17927-PC; CG17927-PD; CG17927-PE; CG17927-PF; CG17927-PG; CG17927-PH; CG17927-PI; CG17927-PK; CG17927-PL; CG17927-PM; CG17927-PN; CG17927-PO; CG17927-PP; CG17927-PQ; CG17927-PR; CG17927-PS; CG17927-PT; CG17927-PU; CG17927-PV; chain; heavy chain; indirect flight muscle (2) RU1; indirect flight muscle (2) RU2; Mhc-PA; Mhc-PB; Mhc-PC; Mhc-PD; Mhc-PE; Mhc-PF; Mhc-PG; Mhc-PH; Mhc-PI; Mhc-PK; Mhc-PL; Mhc-PM; Mhc-PN; Mhc-PO; Mhc-PP; Mhc-PQ; Mhc-PR; Mhc-PS; Mhc-PT; Mhc-PU; Mhc-PV; muscle myosin; muscle myosin heavy chain; muscle myosin II; muscle myosin II heavy chain; muscle myosin-II; muscle specific myosin II; myosin; myosin HC; myosin heavy; Myosin heavy chain, muscle; myosin heavy Chain-PF; myosin heavy-chain; myosin heavy-chain protein; myosin II heavy chain; myosin rod protein; myosin-heavy chain; Not-upheld; samba; sarcomeric myosin heavy-chain; shrunken thorax; Shrunken-thorax; stuckup
Gene Aliases: Bsh; CG17927; chr2L:16765388..16765564; chr2L:16781880..16782040; Dm II; Dmel\CG17927; Dmel_CG17927; DmMHC; DROMHC; DroMII; Ifm(2)2; Ifm(2)RU1; ifm(2)RU2; l(2)36Ae; l(2)k10423; l(2)M66; MHC; Mhc36B; mMHC; MRP; MyHC; Myo; Myo6; Nup; Sht; sMHC; Stp
UniProt ID: (Fruit fly) Q9VJI3
Entrez Gene ID: (Fruit fly) 35007
Molecular Function: G-protein modulator actin binding motor protein actin family cytoskeletal protein cell junction protein cytoskeletal protein enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.