Abnova
Manufacturer Code:PAB8484
Catalog # 89-133-698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Mgp. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Mgp.
For Research Use Only
Protein Aliases: matrix gamma-carboxyglutamate (gla) protein; Matrix Gla protein; Mglap; MGP
Gene Aliases: Mglap; Mgp
UniProt ID: (Mouse) P19788
Entrez Gene ID: (Mouse) 17313
Molecular Function:
annexin
calcium-binding protein
calmodulin
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.