Invitrogen
Manufacturer Code:MA534807
Catalog # PIMA534807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG40-05 |
Immunogen | Recombinant protein of N-terminal human MGEA5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2810009A20Rik 4833427O07Rik 5830447M11Rik AA408215 Beta-hexosaminidase Beta-N-acetylglucosaminidase Beta-N-acetylhexosaminidase Bifunctional protein NCOAT Hexc Hy5 Kiaa0679 MEA5 Meningioma-expressed antigen 5 MGEA5 mKIAA0679 N-acetyl-beta-D-glucosaminidase N-acetyl-beta-glucosaminidase NCOAT Nuclear cytoplasmic O-GlcNAcase and acetyltransferase OGA O-GLCNACASE Protein O-GlcNAcase; Beta-hexosaminidase; Beta-N-acetylglucosaminidase; Beta-N-acetylhexosaminidase; bifunctional protein NCOAT; hyaluronidase in meningioma; Meningioma-expressed antigen 5; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; NCOAT; Nuclear cytoplasmic O-GlcNAcase and acetyltransferase; O-GlcNAcase; OGA; Protein O-GlcNAcase
Gene Aliases: HEXC; KIAA0679; MEA5; MGEA5; NCOAT; OGA
UniProt ID: (Human) Q86WV0
Entrez Gene ID: (Human) 10724
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.