Novus Biologicals
Manufacturer Code:NBP147267
Catalog # NBP147267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 150 and 200 of human Rac GTPase-Activating Protein 1 using the numbering given in entry NP_001119575.1 (GeneID 29127). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CYK4; GTPase activating protein HsCYK-4 ID-GAP KIAA1478 Male germ cell RacGap MGCRACGAP MgcRacGAPrac GTPase-activating protein 1 Rac GTPase activating protein 1; Male germ cell RacGap; MgcRacGAP; Protein CYK4 homolog; Rac GTPase-activating protein 1
Gene Aliases: CYK4; HsCYK-4; ID-GAP; KIAA1478; MGCRACGAP; RACGAP1
UniProt ID: (Human) Q9H0H5
Entrez Gene ID: (Human) 29127
Molecular Function:
G-protein modulator
enzyme modulator
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.