Abnova
Manufacturer Code:MAB18609
Catalog # 89-338-534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S153-5 |
Immunogen | Recombinant protein corresponding to amino acids 370-600 at N-terminus of mouse Mfn2. |
Conjugate | PE/ATTO594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D630023P19Rik; Fzo; HSG protein; hypertension related protein 1; Hypertension-related protein 1; Mitochondrial assembly regulatory factor; Mitofusin-2; Transmembrane GTPase MFN2
Gene Aliases: D630023P19Rik; Fzo; Kiaa0214; Marf; Mfn2
UniProt ID: (Mouse) Q80U63
Entrez Gene ID: (Mouse) 170731
Molecular Function:
hydrolase
nucleotide phosphatase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.