Novus Biologicals
Manufacturer Code:NB10077311
Catalog # NB10077311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 530 and 580 of human methyltransferase like 3 (N6-adenosine-methyltransferase 70 kDa subunit) using the numbering given in entry NP_062826.2 (GeneID 56339). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase EC 2.1.1.62 IME4 M6A methyltransferase like 3 Methyltransferase-like protein 3 MGC4336 mRNA m(6)A methyltransferase MTA70 MT-A70 N6-adenosine-methyltransferase 70 kDa subunit Spo8; hMETTL3; Methyltransferase-like protein 3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit
Gene Aliases: IME4; M6A; METTL3; MT-A70; MTA70; Spo8
UniProt ID: (Human) Q86U44
Entrez Gene ID: (Human) 56339
Molecular Function:
RNA binding protein
RNA methyltransferase
methyltransferase
nucleic acid binding
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.