Novus Biologicals
Manufacturer Code:NBP2822850.1ML
Catalog # NB030214
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human Methyltransferase like 3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase EC 2.1.1.62 IME4 M6A methyltransferase like 3 Methyltransferase-like protein 3 MGC4336 mRNA m(6)A methyltransferase MTA70 MT-A70 N6-adenosine-methyltransferase 70 kDa subunit Spo8; hMETTL3; Methyltransferase-like protein 3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit
Gene Aliases: IME4; M6A; METTL3; MT-A70; MTA70; Spo8
UniProt ID: (Human) Q86U44
Entrez Gene ID: (Human) 56339
Molecular Function: RNA binding protein RNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.