Novus Biologicals
Manufacturer Code:NBP272652B
Catalog # NBP272652B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2C8 |
Immunogen | Full length recombinant protein of human Methylmalonyl Coenzyme A mutase (NP_000246) produced in HEK293T cells. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 5.4.99.2 MCMmethylmalonyl Coenzyme A mutase methylmalonyl CoA mutase Methylmalonyl-CoA isomerase methylmalonyl-CoA mutase mitochondrial; MCM; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; Methylmalonyl-CoA isomerase; Methylmalonyl-CoA mutase, mitochondrial; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase
Gene Aliases: MCM; MMUT; MUT
UniProt ID: (Human) P22033
Entrez Gene ID: (Human) 4594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.