Novus Biologicals
Manufacturer Code:NBP234024
Catalog # NBP234024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SVSMTMNGAVIPVLANFIVTGEEQGVPKEKLTGTIQNDILKEFMVRNTYIFPPEPSMKIIADIFEYTAKHMPKFNSISISGYHMQEAGADAILELAYTLADGLEYSRTGLQAGLTIDEFAPRLSFFWGIG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 5.4.99.2 MCMmethylmalonyl Coenzyme A mutase methylmalonyl CoA mutase Methylmalonyl-CoA isomerase methylmalonyl-CoA mutase mitochondrial; MCM; methylmalonyl CoA mutase; methylmalonyl Coenzyme A mutase; Methylmalonyl-CoA isomerase; Methylmalonyl-CoA mutase, mitochondrial; mutant methylmalonyl CoA mutase; truncated methylmalonyl CoA mutase
Gene Aliases: MCM; MMUT; MUT
UniProt ID: (Human) P22033
Entrez Gene ID: (Human) 4594
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.