Novus Biologicals
Manufacturer Code:NBP290622AF350
Catalog # NB040290
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Mouse Methionine Aminopeptidase 2/METAP2 (O08663 Ala2-Tyr478). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: eIF-2-associated p67 homolog; eIF-2-associated p67 homolog Initiation factor 2-associated 67 kDa glycoprotein MAP 2 MAP2 MetAP 2 methionine aminopeptidase 2 methionyl aminopeptidase 2 MNPEP p67 p67EC 3.4.11.18 P67EIF2 Peptidase M 2; Initiation factor 2-associated 67 kDa glycoprotein; MAP 2; Methionine aminopeptidase 2; p67; Peptidase M; peptidase M 2; testicular tissue protein Li 17
Gene Aliases: MAP2; METAP2; MNPEP; P67EIF2
UniProt ID: (Human) P50579
Entrez Gene ID: (Human) 10988
Molecular Function:
hydrolase
metalloprotease
nucleic acid binding
protease
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.