R&D Systems
Manufacturer Code:IC9219R100UG
Catalog # IC9219R100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2030b |
Immunogen | <em>E. coli</em>-dervived recombinant human MESP1 Met1-Gln85 Accession No. Q9BRJ9 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8°C as supplied. |
Rabbit Monoclonal Antibody
Protein Aliases: bHLHc5; BHLHC5 bHLHc5MGC10676 Class C basic helix-loop-helix protein 5 mesoderm posterior 1 homolog (mouse) mesoderm posterior protein 1; Class C basic helix-loop-helix protein 5; mesoderm posterior 1 homolog; mesoderm posterior basic helix-loop-helix transcription factor 1; Mesoderm posterior protein 1
Gene Aliases: BHLHC5; MESP1
UniProt ID: (Human) Q9BRJ9
Entrez Gene ID: (Human) 55897
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.