Novus Biologicals
Manufacturer Code:NBP27975220UG
Catalog # NB010522
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ENAH/1988 |
Immunogen | Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ENA enabled homolog (Drosophila) FLJ10773 MENANDPP1 protein enabled homolog; mammalian enabled variant 11a; mammalian enabled variant pan; Protein enabled homolog
Gene Aliases: ENA; ENAH; MENA; NDPP1
UniProt ID: (Human) Q8N8S7
Entrez Gene ID: (Human) 55740
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.