Novus Biologicals
Manufacturer Code:NLS932R
Catalog # NLS932R
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286]. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FGQTL2 Mel 1b melatonin receptor Mel1b receptor MEL-1B-R melatonin receptor 1B melatonin receptor MEL1B melatonin receptor type 1B MT2; Mel-1B-R; mel1b receptor; melatonin receptor 1B variant b; melatonin receptor MEL1B; Melatonin receptor type 1B
Gene Aliases: FGQTL2; MEL-1B-R; MT2; MTNR1B
UniProt ID: (Human) P49286
Entrez Gene ID: (Human) 4544
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.