Novus Biologicals
Manufacturer Code:NBP2841590.1ML
Catalog # NB032087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Melatonin R1A/MT1/MTNR1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mel-1A-R; mel1a receptor; Mel1a receptor MEL-1A-R melatonin receptor 1A melatonin receptor type 1A MT1; Melatonin receptor type 1A
Gene Aliases: MEL-1A-R; MT1; MTNR1A
UniProt ID: (Human) P48039
Entrez Gene ID: (Human) 4543
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.