R&D Systems
Manufacturer Code:MAB8175SP
Catalog # MAB8175SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 893438 |
Immunogen | HEK293 human embryonic kidney cell line transfected with human Melanotransferrin/CD228 Met1-Gly711 Accession # P08582 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5 CD228 CD228 antigen FLJ38863 MAP97 MAP97Melanoma-associated antigen p97 melanotransferrin membrane-bound transferrin-like protein MFI2 MGC4856 MTF MTF1 p97; CD228; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; Melanotransferrin; membrane-bound transferrin-like protein
Gene Aliases: CD228; MAP97; MELTF; MFI2; MTf; MTF1
UniProt ID: (Human) P08582
Entrez Gene ID: (Human) 4241
Molecular Function:
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.