R&D Systems
Manufacturer Code:FAB8205V100UG
Catalog # FAB8205V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 821012 |
Immunogen | NS0 mouse myeloma cell line transfected with human Melanocortin-5 R/MC5R Met1-Asp325 Accession # P33032 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: MC-2; MC2 MC-2 MC5 MC5R MC5-R melanocortin 5 receptor melanocortin receptor 5 melanocortin-5 receptor Melanocortin5R Melanocortin-5R; MC5-R; Melanocortin receptor 5; melanocortin-5 receptor
Gene Aliases: MC2; MC5R
UniProt ID: (Human) P33032
Entrez Gene ID: (Human) 4161
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.