Novus Biologicals
Manufacturer Code:NBP2841580.1ML
Catalog # NB032086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Melanocortin-2 R/MC2R |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACTH receptor; ACTH receptor ACTH-R ACTHRcorticotropin receptor adrenocorticotropic hormone receptor Adrenocorticotropin receptor MC2 receptor MC2-R melanocortin 2 receptor (adrenocorticotropic hormone) Melanocortin receptor 2 MGC125798; Adrenocorticotropic hormone receptor; Adrenocorticotropin receptor; corticotropin receptor; MC2 receptor; MC2-R; melanocortin 2 receptor (adrenocorticotropic hormone); Melanocortin receptor 2
Gene Aliases: ACTHR; MC2R
UniProt ID: (Human) Q01718
Entrez Gene ID: (Human) 4158
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.