Abnova
Manufacturer Code:PAB8968
Catalog # 89-134-056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 194-209 of fruit fly Med. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Med.
For Research Use Only
Protein Aliases: anon-EST:Posey121; CG1775; CG1775-PA; CG1775-PB; CG1775-PC; class II Smad; dSmad4; E(zen)3; l(3)11m-254; l(3)12m-137; l(3)SG36; l(3)SG70; l(3)XIIm137; LD22279p; lethal(3)SG36; lethal(3)SG70; Med-PA; Med-PB; Med-PC; medea; media
Gene Aliases: anon-EST:Posey121; CG1775; Dmel\CG1775; Dmel_CG1775; dSmad4; E(zen)3; l(3)11m-254; l(3)12m-137; l(3)SG36; l(3)SG70; l(3)XIIm137; MED; medea; SMAD4
Entrez Gene ID: (Fruit fly) 43725
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.