Abnova
Manufacturer Code:PAB15647
Catalog # 89-117-714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at internal region of mouse Mcpt4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised aganist synthetic peptide of Mcpt4.
For Research Use Only
Protein Aliases: Mast cell protease 4; Mcp-4; MMCP-4; MMCP-4A; MMCP-4B; MSMCP; Myonase; Serosal mast cell protease
Gene Aliases: Mcp-4; Mcp4; Mcpt4; MMCP-4; MMCP-4A; MMCP-4B
UniProt ID: (Mouse) P21812
Entrez Gene ID: (Mouse) 17227
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.