R&D Systems
Manufacturer Code:MAB8281
Catalog # MAB8281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 602901 |
Immunogen | <EM>E. coli</EM>-derived recombinant human Mcl-1 Met231-Arg350 Accession # Q07820 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Bcl-2-like protein 3; Bcl-2-related protein EAT/mcl1; Bcl2-L-3; bcl2-L-3 BCL2L3 BCL2L3MGC104264 Bcl-2-like protein 3 Bcl-2-related protein EAT/mcl1 EAT induced myeloid leukemia cell differentiation protein Mcl-1 Mcl1 Mcl-1 mcl1/EAT MCL1-ES MCL1L MCL1S MGC1839 myeloid cell leukemia ES myeloid cell leukemia sequence 1 (BCL2-related) TM; Induced myeloid leukemia cell differentiation protein Mcl-1; mcl1/EAT; myeloid cell leukemia 1; myeloid cell leukemia ES; myeloid cell leukemia sequence 1 (BCL2-related)
Gene Aliases: bcl2-L-3; BCL2L3; EAT; Mcl-1; MCL1; MCL1-ES; mcl1/EAT; MCL1L; MCL1S; TM
UniProt ID: (Human) Q07820
Entrez Gene ID: (Human) 4170
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.