Abnova
Manufacturer Code:PAB27605
Catalog # 89-129-116
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region of mouse Mboat4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Mboat4.
For Research Use Only
Protein Aliases: Ghrelin O-acyltransferase; Gm171; GOAT; Membrane-bound O-acyltransferase domain-containing protein 4; O-acyltransferase domain-containing protein 4
Gene Aliases: Gm171; GOAT; Mboat4
UniProt ID: (Mouse) P0C7A3
Entrez Gene ID: (Mouse) 234155
Molecular Function:
acetyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.