Novus Biologicals
Manufacturer Code:NBP233726
Catalog # NBP233726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: INLCDMCVILSANQNNIDDTSLQDKECILASLNIKSMQFDDSIGVLQANSQGFTPPGMDRSSPDNSPVHGMLRQPSITTGVNIPIITELVNDTNVQFLDQDDDDDPDTELYLTQP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: bA205K10.1 BK channel BKCA alpha BKCA alpha subunit BKTM calcium-activated potassium channel subunit alpha-1 DKFZp686K1437 hSlo k(VCA)alpha KCNMA Maxi K channel MaxiK MGC71881 potassium large conductance calcium-activated channel subfamily M alphamember 1 SAKCA Slo homolog Slo1 SLO-ALPHA Slowpoke homolog stretch-activated Kca channel subfamily M subunit alpha-1; big potassium channel alpha subunit; BK channel; BK channel alpha subunit; BKCA alpha; BKCA alpha subunit; Calcium-activated potassium channel subunit alpha-1; Calcium-activated potassium channel, subfamily M subunit alpha-1; K(VCA)alpha; KCa1.1; Maxi K channel; maxi-K channel HSLO; MaxiK; potassium channel, calcium activated large conductance subfamily M alpha, member 1; potassium large conductance calcium-activated channel, subfamily M, alpha member 1; Slo homolog; Slo-alpha; Slo1; Slowpoke homolog; stretch-activated Kca channel
Gene Aliases: bA205K10.1; BKTM; hSlo; KCa1.1; KCNMA; KCNMA1; MaxiK; mSLO1; SAKCA; SLO; SLO-ALPHA; SLO1
UniProt ID: (Human) Q12791
Entrez Gene ID: (Human) 3778
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.