Novus Biologicals
Manufacturer Code:NBP160091
Catalog # NBP160091
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MAS1(MAS1 oncogene) The peptide sequence was selected from the middle region of MAS1. Peptide sequence VIIFIAILSFLVFTPLMLVSSTILVVKIRKNTWASHSSKLYIVIMVTIII. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MAS MAS1 oncogene MGC119966 proto-oncogene Mas; Mas-related G protein-coupled receptor A; MAS1 oncogene; Proto-oncogene Mas
Gene Aliases: MAS; MAS1; MGRA
UniProt ID: (Human) P04201
Entrez Gene ID: (Human) 4142
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.