Abnova
Manufacturer Code:MAB1678
Catalog # 89-110-572
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | KT51 |
Immunogen | Recombinant GST fusion protein corresponding to amino acids 125-268 of mouse Mapre1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against recombinant Mapre1.
For Research Use Only
Protein Aliases: 5530600P05Rik; adenomatosis polyposis coli binding protein Eb1; AI462499; AI504412; APC-binding protein EB1; AW260097; BIM1p; D2Ertd459e; EB1; End-binding protein 1; Microtubule-associated protein RP/EB family member 1
Gene Aliases: 5530600P05Rik; AI462499; AI504412; AW260097; BIM1p; D2Ertd459e; Eb1; Mapre1
UniProt ID: (Mouse) Q61166
Entrez Gene ID: (Mouse) 13589
Molecular Function: cytoskeletal protein microtubule family cytoskeletal protein non-motor microtubule binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.