Abnova
Manufacturer Code:MAB18648
Catalog # 89-338-573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S135-37 |
Immunogen | Recombinant protein corresponding to amino acids 226-421 of mouse Mapk8ip2. |
Conjugate | ATTO655 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3230402N03Rik; AI847694; AW049958; C-Jun-amino-terminal kinase-interacting protein 2; IB-2; Islet-brain-2; JIP-2; Jip2; JNK MAP kinase scaffold protein 2; JNK-interacting protein 2; mitogen activated protein kinase 8 interacting protein 2; Mitogen-activated protein kinase 8-interacting protein 2; R75148
Gene Aliases: 3230402N03Rik; AI847694; AW049958; IB2; Jip2; Mapk8ip2; R75148
UniProt ID: (Mouse) Q9ERE9
Entrez Gene ID: (Mouse) 60597
Molecular Function:
enzyme modulator
kinase activator
kinase modulator
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.