Novus Biologicals
Manufacturer Code:NBP272584F
Catalog # NBP272584F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G7 |
Immunogen | Full length recombinant protein of human Mannose Phosphate Isomerase (NP_002426) produced in HEK293T cells. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: CDG1B EC 5.3.1 EC 5.3.1.8 FLJ39201 mannose phosphate isomerase mannose-6- phosphate isomerase mannose-6-phosphate isomerase Phosphohexomutase Phosphomannose isomerase phosphomannose isomerase 1 PMI PMI1phosphohexomutase; Mannose-6-phosphate isomerase; Phosphohexomutase; Phosphomannose isomerase; phosphomannose isomerase 1; PMI
Gene Aliases: CDG1B; MPI; PMI; PMI1
UniProt ID: (Human) P34949
Entrez Gene ID: (Human) 4351
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.