R&D Systems
Manufacturer Code:MAB3809
Catalog # MAB3809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 381923 |
Immunogen | <EM>E. coli</EM>-derived recombinant human MafK Thr2-Ser156 Accession # O60675 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: basic-leucine zipper transcription factor MafK; Erythroid transcription factor NF-E2 p18 subunit; FLJ32205 MGC71717 NFE2U transcription factor MafK ubiquitous (p18) v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein K v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family protein K v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian); nuclear factor erythroid-2, ubiquitous (p18); Transcription factor MafK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Aliases: MAFK; NFE2U; P18
UniProt ID: (Human) O60675
Entrez Gene ID: (Human) 7975
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.