Novus Biologicals
Manufacturer Code:NBP256366
Catalog # NBP256366
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:ITIVKSTELSSTSVPFSAAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic-leucine zipper transcription factor MafK; Erythroid transcription factor NF-E2 p18 subunit; FLJ32205 MGC71717 transcription factor MafK ubiquitous (p18) v-maf avian musculoaponeurotic fibrosarcoma oncogene family protein K v-maf musculoaponeurotic fibrosarcoma (avian) oncogene family protein K v-maf musculoaponeurotic fibrosarcoma oncogene homolog K (avian); nuclear factor erythroid-2, ubiquitous (p18); Transcription factor MafK; v-maf avian musculoaponeurotic fibrosarcoma oncogene family, protein K; v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K; v-maf musculoaponeurotic fibrosarcoma oncogene homolog K
Gene Aliases: MAFK; NFE2U; P18
UniProt ID: (Human) O60675
Entrez Gene ID: (Human) 7975
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.